Learn More
KGW Isotherm™ KGW Isotherm
Brand:  KGW Isotherm™ 17111
8753.06 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15311271
  • 9325.00 DKK / Each
Only null left
Estimated Shipment: 06-08-2025
Specifications
200mL Condensate, 1.2L Coolant, stainless Steel
1 Pack
Cold Trap
Certificates
Promotions
Promotions
Product Content Correction
Product Title
KGW Isotherm™ KGW Isotherm > Material: Stainless Steel Casing