Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Available in natural (clear) or choice of four colors
Brand:  DWK Life Sciences 73662-13
 View more versions of this product
Product Code. 10389354
  • 1170.00 DKK / Pack of 1000
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Push Cap
Polypropylene
Non-sterile
Reusable
Autoclavable
Red
13 mm Plain-end Culture Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Kimble™ Kim-Kap™ Closures > For O.D.: 13 mm; Color: Red