Learn More
DWK Life Sciences Kimble™ Kontes™ Adsorption/Ion-Exchange Columns
With or without fritted disc
Brand: DWK Life Sciences 420320-0245
Description
- Borosilicate glass
- Sealed-in coarse porosity (40 to 60μm) fritted disc supports packing in K420320 Series columns
- Use glass wool to support packing in K420300 Series columns (packing not included)
Specifications
Many separations in adsorption and ion exchange chromatography | |
Adsorption/Ion Exchange Chromatography Column | |
250 mL | |
Borosilicate Glass |
25 mm | |
40 to 60 μm | |
500 mm | |
Fritted disc |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.