Learn More
DWK Life Sciences Kimble™ Kontes™ FlexColumn Economy Columns Flow Adapters
Convert standard FlexColumn™ columns to adjustable bed volume columns
Brand:  DWK Life Sciences 420415-2500
 View more versions of this product
Product Code. 11708242
  • 3230.00 DKK / Each
Estimated Shipment: 29-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Includes
  • PTFE eluent cannula
  • Polyacetal body
  • Viton™ O-ring seal
  • One replacement 20μm polyethylene bed support
Order Info
Additional bed supports can be ordered in packs of 10 (see K420415-1001, -1501, -2501).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Concentration and pH gradients, reverse flow and gravity flow separation
Flow Control Adapter
Polyacetal
0.062 in. O.D. FEP PTFE tubing, Viton™ FKM O-ring seal, replacement 20um porosity HDPE bed support
25 mm I.D. FlexColumn
140 mm
HDPE
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Kimble™ Kontes™ FlexColumn Economy Columns Flow Adapters > Fits 2.5cm I.D.; Functional length 14cm