Learn More
DWK Life Sciences Kimble™ Kontes™ HPLC Economy 3-Hole Ultra-Ware Cap Systems
Click to view available options
Capacity (Metric):
1 L
5 L
Description
- Made of type I, Class A borosilicate glass with a plastic safety coating
- Cap body is made of PTFE
- Supplied with TFE/propylene o-ring and polypropylene screw collar
- Connecting threads on top of cap use standard 1/4-28 flangeless fittings
- Two ports have 1/8 in. through holes, third port has 1/16 in. through hole
Specifications
Specifications
Capacity (Metric) | 1 L |
Description | Economy Three Hole Cap System with 1 Liter Reservoir |
Includes | TFE/Propylene o-ring and Polypropylene screw collar, 1L reservoir |
Material | PTFE |
No. per Case | 1 Cs. |
For Use With (Equipment) | Bottle |