Learn More
Kimberly-Clark Professional™ Kimtech™ M3 Pleat-Style Facemasks with Ties
Pleated style face mask with ties providing excellent bacterial and particle filtration to avoid contamination. Low-lint polyethylene film outer layer. Kimtech™ M3 Pleat-Style Facemasks with Ties are latex and silicone free. Suitable for use in ISO 3 or higher cleanrooms.
Brand: Kimberly-Clark Professional™ 62466
1294.54 DKK valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
- Excellent bacterial and particle filtration to avoid contamination.
- Low-lint polyethylene film outer layer.
- Latex and silicone free.
- Suitable for use in ISO 3 or higher cleanrooms.
Specifications
Cleanroom Face Mask | |
Ties | |
Universal | |
Pleated | |
Single-use | |
Soft Ties | |
62466 |
White | |
Polyethylene | |
Non-sterile | |
ISO 14001, ISO 9001, CE CAT I, Industrial Use Only | |
3.0 micron | |
Yes |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02