Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
404.00 DKK - 5830.00 DKK
Products 14
Product Code Brand Product Type For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Product Type For Use With (Equipment) Price Quantity & Availability  
16542452
View Documents Promotion Details
Thermo Scientific™
97002570
Tip Comb KingFisher Apex
5270.00 DKK
Pack of 100
10373480
View Documents Promotion Details
Thermo Scientific™
95040450
96 Deep Well plate, v-bottom KF Duo Prime, Flex and Presto
1848.00 DKK
Pack of 50
10329827
View Documents Promotion Details
Thermo Scientific™
95040460
Sterile 96 Deep Well Plate KF Duo Prime, Flex and Presto
2330.00 DKK
Pack of 50
11744978
View Documents Promotion Details
Thermo Scientific™
97002534
Tip Comb KingFisher Flex and Presto
4765.00 DKK
Pack of 100
15698465
View Documents Promotion Details
Thermo Scientific™
97002820
Tip Comb and Plate KingFisher Flex and Presto
5650.00 DKK
Pack of 50
12693116
View Documents Promotion Details
Thermo Scientific™
97003500
Tip Comb KingFisher Duo Prime only
664.00 DKK
Pack of 50
12633116
View Documents Promotion Details
Thermo Scientific™
97003520
Elution Strip KingFisher Duo Prime only
532.00 DKK
Pack of 40
15357317
View Documents Promotion Details
Thermo Scientific™
97003540
Elution Strip Cap KingFisher Duo Prime only
404.00 DKK
Pack of 40
11680243
View Documents Promotion Details
Thermo Scientific™
97003530
Combi Pack KingFisher Duo Prime only
742.00 DKK
Each
16882406
View Documents
Thermo Scientific™
95040455
96-well Deep Well Plate KingFisher Flex, Duo Prime This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17120069
View Documents Promotion Details
Thermo Scientific™
95040450B
96-well Deep Well Plate KingFisher Apex
2095.00 DKK
Pack of 50
17170069
View Documents Promotion Details
Thermo Scientific™
97002534B
Barcoded Tip Comb KingFisher Apex
5020.00 DKK
Pack of 100
17130069
View Documents Promotion Details
Thermo Scientific™
95040460B
Sterile 96 Deep Well Plate KingFisher Apex
2440.00 DKK
Pack of 50
17110079
View Documents Promotion Details
Thermo Scientific™
97002820B
Tip Comb and Plate KingFisher Apex
5830.00 DKK
Pack of 50
Description
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ KingFisher™ Plastics for 96 deep-well format