Learn More
Thermo Scientific™ KingFisher™ Flex Purification System
Prepare samples efficiently from a variety of materials with Thermo Scientific KingFisher Flex Purification Systems, which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source.
Brand:  Thermo Scientific™ 24074411
 View more versions of this product
Product Code. 12501758
  • 36300.00 DKK / Each
Description
Prepare samples efficiently from a variety of materials with Thermo Scientific KingFisher Flex Purification Systems, which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein or cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproduciblity and quality.
Find a flexible solution for different types of sample processing needs. With 24 configurations, customers can raise the processing volume up to 5mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.
• Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
• High throughput system processes up to 96 samples in less than 20 minutes.
• Expands sample volume by 10-fold with the 24-well format.
• Open and flexible system lets the customer use anymagnetic particle-based kit tomeet the application demands.
• Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation andmodification.
• Ready-made protocols for different type of applications are available.
Recommended for:
• Genomics and proteomics
• Target identification
• Veterinary assays
• Biomarker discovery
• Quality control
• Forensics
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96 PCR head and heating block
20 min.
Videos
  • Play Video
    Play
    Mute
    Current Time 0:00
    /
    Duration Time 0:00
    Loaded: 0%
    Progress: 0%
    Stream TypeLIVE
    Remaining Time -0:00
    Fullscreen
    Share
    Close
    Share Video:
    Thermo Scientific KingFisher Flex Live Demo
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ KingFisher™ Flex Purification System >
For Laboratory Use