Learn More
Thermo Scientific™ KingFisher Plastic Consumables for SARS-CoV-2 Testing
These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.
Brand: Thermo Scientific™ A48438
Description
These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.
Optimize performance using Thermo Scientific KingFisher Systems Consumables, designed specifically for use for COVID-19 testing with these instruments. KingFisher instruments utilize disposable plastics made of polypropylene. The plastics, tip combs and microplates, are ideal for magnetic particle processing because of their low binding affinity for biomolecules. The special design enables excellent recovery of magnetic beads. Specifically tested with the TaqPath COVID-19 Combo Kit and MagMAX Viral/Pathogen Nucleic Acid Isolation Kit.For TaqPath COVID-19 testing with MagMAX Viral/Pathogen Nucleic Acid Isolation Kit
Specifications
96-well Plate | |
KingFisher Deep Well 96 Tip Comb, 100 PCS | |
100/Pk. |
96 Samples | |
KingFisher Flex and Presto |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.