Learn More
LNI Swissgas Kit Service for NG SIRIO 10 L
Kit service 16000 h for NG SIRIO 10 L.
Brand: LNI Swissgas 6930.00.196
This item is not returnable.
View return policy
Description
- To be carried out by trained service engineer every 16000 hours or 4 years.
Specifications
PM Kit |
Product Content Correction
Product Title
LNI Swissgas Kit Service for NG SIRIO 10 L > LNI Swissgas Kit Service
Spot an opportunity for improvement?Share a Content Correction