Learn More
Kjeldahl Boric Acid Solution, Pure, Solution 4% Alkaline with Indicator, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
Specifications
Specifications
Color | Red |
Quantity | 1 L |
Packaging | Plastic Container |
Physical Form | Liquid |
MDL Number | 11337 |
Product Content Correction
Product Title
Kjeldahl Boric Acid Solution, Pure, Solution 4% Alkaline with Indicator, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction