Learn More
Kimberly-Clark™ KleenGuard™ Jackson Safety™ V20 Purity Eyewear
Allows you to provide stylish, yet economical eye protection for yourself and your team. KleenGuard™ Jackson Safety™ V20 Purity Eyewear features padded temples for enhanced comfort.
447.00 DKK
Specifications
Certifications/Compliance | EN166:2001 1F for safety, EN166:2001 level F for impact resistance and EN166:2001 class 1 for optical clarity. |
---|---|
Lens Tint | Clear |
Product Type | Safety Eyewear |
No. per Pack | 12 Pk. |
Material | Plastic |
Description
- Complies with: EN166:2001 1F for safety
- Complies with: EN166:2001 level F for impact resistance
- Complies with: EN166:2001 class 1 for optical clarity
- 99% UVA/UVB protection
- Comfortable to wear for long periods
- Economical eye protection for workers and visitors
Specifications
EN166:2001 1F for safety, EN166:2001 level F for impact resistance and EN166:2001 class 1 for optical clarity. | |
Safety Eyewear | |
Plastic | |
Polycarbonate | |
UV400 |
Clear | |
12 Pk. | |
Reusable | |
Plastic | |
Frameless |