missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLK7 293T Cell Overexpression Lysate 1 (Denatured), Abnova
Brand: Abnova H00005650-T01.100uL
Additional Details : Weight : 0.00010kg
Description
Western Blotting
Specifications
Western Blot | |
KLK7 293T Cell Transient Overexpression Lysate(Denatured) | |
27.94 | |
pCMV-KLK7 full-length | |
100 μL | |
Human |
293T | |
BC032005 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-KLK7 antibody (H00005650-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |