Learn More
DWK Life Sciences Kimble™ Kontes™ Disposaflex™ Lower Column Fitting
For Disposaflex Disposable Polypropylene Column Set
Brand:  DWK Life Sciences 420168-0000
Product Code. 11941027
  • 918.00 DKK / Pack of 100
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Includes recess to hold polyethylene bed support or glass wool plug
  • Polypropylene
  • Pack of 100
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene