Learn More
DWK Life Sciences Kimble™ Kontes™ Disposaflex™ Stopcocks
For Disposaflex™ Chromatography Columns
3360.00 DKK
Specifications
Type Valve
For Use With Disposaflex Chromatography Column
View More Specs
Products 1
Product Code Brand Material Price Quantity & Availability  
Product Code Brand Material Price Quantity & Availability  
11806121
View Documents
DWK Life Sciences
420163-0000
Polycarbonate stopcock; 1-Way F-Luer to M-Luer
3360.00 DKK
Pack of 50
Only null left
Estimated Shipment: 05-11-2025
Add to basket
Description
Description
For Disposaflex™ Chromatography Columns
  • Polycarbonate Body
  • HDPE Plug
Specifications
Specifications
Valve
Disposaflex Chromatography Column
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Kimble™ Kontes™ Disposaflex™ Stopcocks