Learn More
DWK Life Sciences Kimble™ Kontes™ Reusable Pellet Pestles™ Tissue Grinders
For use with 15mL micro centrifuge tubes
Brand:  DWK Life Sciences 749515-1500
Product Code. 11805731
  • 1950.00 DKK / Each
Only null left
Estimated Shipment: 11-09-2025
Description
For use with 15mL micro centrifuge tubes
  • Autoclavable CTFE/stainless-steel
  • Specially designed to match microtubes
  • Size: 15mL
  • Overall Pestle Length: 21cm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
15mL Micro Tubes
8.26 in.
CTFE/Stainless Steel
15 mL
Cs. of 1
210mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Reusable Pellet Pestles™ Tissue Grinders > Pellet Pestle, Reusable; 15mL