Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
8100.00 DKK - 11335.00 DKK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
11858482
|
DWK Life Sciences
953825-0000 |
300 mL (Funnel), 1000 mL (Flask) |
8100.00 DKK
Each |
Estimated Shipment: 27-03-2026
Log in to see stock. |
||||||
11868522
|
DWK Life Sciences
953835-0000 |
500 mL (Funnel), 2000 mL (Flask) |
10515.00 DKK
Each |
Estimated Shipment: 27-03-2026
Log in to see stock. |
||||||
11719055
|
DWK Life Sciences
953845-0000 |
1000 mL (Funnel), 4000 mL (Flask) |
11335.00 DKK
Each |
Estimated Shipment: 27-03-2026
Log in to see stock. |
||||||
Description
Recommended for routine filtration analysis of corrosive liquids and the removal of particulates from HPLC solvents
- The ground joint connection eliminates the phthalate contamination that can occur when using silicone or neoprene stoppers
- The support base has a 40-60 micron porosity glass frit and an integral vacuum connection that is located above the drip tip to prevent contamination of the vacuum line with filtrate droplets
- Each apparatus is supplied with a funnel, an anodized aluminum clamp, a 47 mm fritted glass support base and a filtration flask
- Joints are 40/35
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Glass | |
Microfiltration Assembly |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly with Fritted Glass Support and Ground Joint (47mm)