Learn More
IKA™ KS 260 Control NOL Flat Shaker
GREENER_CHOICE
Compact, flat shaker with ideal swivel motion.
Brand:  IKA™ 0004278400
Product Code. 15895931
  • 27595.00 DKK / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Electronic adjustment of speed and timer
  • Digital display makes it possible to read the speed, timer function and operating modes
  • Electronic time switching clock
  • All functions can be controlled and documented with labworldsoft™ software
  • Special version with reverse rotating direction on request
  • Wide range of attachment combinations makes it possible to use almost all shapes and sizes of vessels
  • Attachments are not included, please order separately