Learn More
L 888607 Racemate, MedChemExpress
MedChemExpress L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.
Brand: Medchem Express HY-111271A
Description
L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.Specifications
L 888607 Racemate | |
Light Yellow | |
C19H15ClFNO2S | |
DMSO : 6 mg/mL (15.96 mM; Need ultrasonic) | |
375.84 | |
99.44% | |
Solid |
1030017-51-6 | |
Research | |
10 mg | |
O=C(O)CC1CCN2C1=C(SC3=CC=C(Cl)C=C3)C4=C2C=C(F)C=C4 | |
375.84 | |
Research | |
Neuroscience-Neuromodulation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only