Learn More
L 888607 Racemate, MedChemExpress
MedChemExpress L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.
Brand:  Medchem Express HY-111271A
Encompass_Preferred
 View more versions of this product
Product Code. 16487137
  • 3333.58 DKK / 5mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
L 888607 Racemate is a selective prostaglandin D2 receptor subtype 1 (DP1) antagonist, with Kis of 132 nM and 17 nM for DP1 and thromboxane A2 receptor (TP), respectively.
Specifications
Specifications
L 888607 Racemate
Light Yellow
C19H15ClFNO2S
DMSO : 6 mg/mL (15.96 mM; Need ultrasonic)
375.84
99.44%
Solid
1030017-51-6
Research
5 mg
O=C(O)CC1CCN2C1=C(SC3=CC=C(Cl)C=C3)C4=C2C=C(F)C=C4
375.84
Research
Neuroscience-Neuromodulation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
L 888607 Racemate, MedChemExpress