Learn More
Thermo Scientific Chemicals L-Cysteine methyl ester hydrochloride
459.05 DKK - 2243.07 DKK
Chemical Identifiers
CAS | 18598-63-5 |
---|---|
Molecular Formula | C4H10ClNO2S |
Molecular Weight (g/mol) | 171.639 |
MDL Number | MFCD00038998 |
InChI Key | WHOHXJZQBJXAKL-DFWYDOINSA-N |
Synonym | l-cysteine methyl ester hydrochloride, methyl l-cysteinate hydrochloride, h-cys-ome.hcl, l-cystein methyl ester hydrochloride, acdrile, actiol, mecysteine hydrochloride, cysteine methyl ester hydrochloride, r-methyl 2-amino-3-mercaptopropanoate hydrochloride |
PubChem CID | 2733208 |
IUPAC Name | methyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride |
SMILES | COC(=O)C(CS)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15486379
|
Thermo Scientific Chemicals
J64436.14 |
25 g |
459.05 DKK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15496379
|
Thermo Scientific Chemicals
J64436.22 |
100 g |
1304.11 DKK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15406389
|
Thermo Scientific Chemicals
J64436.30 |
250 g |
2243.07 DKK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Cysteine methyl ester hydrochloride is a biochemical shown to inhibit binding of ethynylestradiol metabolites to protein and nucleic acids. It used in the synthesis of peptides, Preparation of thiazolidine-4-carboxylates with carbonyl compounds.
Solubility
Soluble in water
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
18598-63-5 | |
171.639 | |
WHOHXJZQBJXAKL-DFWYDOINSA-N | |
2733208 | |
COC(=O)C(CS)N.Cl |
C4H10ClNO2S | |
MFCD00038998 | |
l-cysteine methyl ester hydrochloride, methyl l-cysteinate hydrochloride, h-cys-ome.hcl, l-cystein methyl ester hydrochloride, acdrile, actiol, mecysteine hydrochloride, cysteine methyl ester hydrochloride, r-methyl 2-amino-3-mercaptopropanoate hydrochloride | |
methyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride |
Specifications
142°C (decomposition) | |
White | |
MFCD00038998 | |
14,5785 | |
Soluble in water | |
COC(=O)C(CS)N.Cl | |
171.639 | |
171.65 | |
L-Cysteine methyl ester hydrochloride |
18598-63-5 | |
C4H10ClNO2S | |
3685824 | |
l-cysteine methyl ester hydrochloride, methyl l-cysteinate hydrochloride, h-cys-ome.hcl, l-cystein methyl ester hydrochloride, acdrile, actiol, mecysteine hydrochloride, cysteine methyl ester hydrochloride, r-methyl 2-amino-3-mercaptopropanoate hydrochloride | |
WHOHXJZQBJXAKL-DFWYDOINSA-N | |
methyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride | |
2733208 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 242-435-0
missing translation for 'rtecsNumber' : HA2460000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only