Learn More
L-Methionine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals
437.04 DKK - 952.38 DKK
Chemical Identifiers
CAS | 2899-36-7 |
---|---|
Molecular Formula | C7H15NO2S·HCl |
Molecular Weight (g/mol) | 213.73 |
InChI Key | KPWCQEUBJAIORR-RGMNGODLSA-N |
Synonym | l-methionine ethyl ester hydrochloride, ethyl l-methionate hydrochloride, h-met-oet.hcl, ethyll-methionatehydrochloride, l-methionine, ethyl ester, hydrochloride, h-met-oet hcl, ethyl l-methionate hcl, ethyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-oet?cl, pubchem14924 |
PubChem CID | 3083785 |
IUPAC Name | ethyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride |
SMILES | CCOC(=O)C(CCSC)N.Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2899-36-7 | |
213.73 | |
l-methionine ethyl ester hydrochloride, ethyl l-methionate hydrochloride, h-met-oet.hcl, ethyll-methionatehydrochloride, l-methionine, ethyl ester, hydrochloride, h-met-oet hcl, ethyl l-methionate hcl, ethyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-oet?cl, pubchem14924 | |
ethyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride |
C7H15NO2S·HCl | |
KPWCQEUBJAIORR-RGMNGODLSA-N | |
3083785 | |
CCOC(=O)C(CCSC)N.Cl |
Specifications
2899-36-7 | |
98.5% min. (Argentometry) | |
C7H15NO2S·HCl | |
+ 21.00 (25.00°C c=2,EtOH) | |
l-methionine ethyl ester hydrochloride, ethyl l-methionate hydrochloride, h-met-oet.hcl, ethyll-methionatehydrochloride, l-methionine, ethyl ester, hydrochloride, h-met-oet hcl, ethyl l-methionate hcl, ethyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-met-oet?cl, pubchem14924 | |
CCOC(=O)C(CCSC)N.Cl | |
213.73 | |
213.73 | |
L-Methionine ethyl ester hydrochloride, 99% |
82.0°C to 92.0°C | |
Glass bottle | |
CH3SCH2CH2CH(NH2)CO2C2H5·HCl | |
+ 21.00 | |
KPWCQEUBJAIORR-RGMNGODLSA-N | |
ethyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride | |
3083785 | |
99% |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 220-787-6
- L-Methionine ethyl ester hydrochloride, 99%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.