Learn More
Microspec™ L Shaped Spreader
Sterile L shaped inoculating spreaders
Brand: Microspec™ E7G
This item is not returnable.
View return policy
Description
- L Shaped Spreaders
- 39 mm x 140 mm
- Microseal Pouch
- Sterile EtO
Specifications
Single-use | |
Individually Wrapped in Microseal Pouches | |
1.5 x 16.1 in. | |
Plastic | |
Sterile |
Not Autoclavable | |
250 | |
39 x 410 mm | |
Inoculating Spreader |
Product Content Correction
Product Title
Microspec™ L Shaped Spreader > Packaging: Individually Wrapped in Microseal Pouches
Spot an opportunity for improvement?Share a Content Correction