missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Valinol, 97%

Product Code. 11317686
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 2026-48-4 | C5H13NO | 103.17 g/mol

L-(+)-Valinol reacts with aldehydes and nitriles to form imines and oxazolines, respectively, for asymmetric synthesis. It is also used for preparation of 4-isopropyloxazolidinones, employed in stereocontrolled aldol and alkylation reactions, synthesis of chiral 2-thiazolidinethiones for aldol reactions, asymmetric alkylation of condensation product with keto acids, asymmetric reactions of its amidines and imines and oxidation of N-protected derivative to the aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Valinol reacts with aldehydes and nitriles to form imines and oxazolines, respectively, for asymmetric synthesis. It is also used for preparation of 4-isopropyloxazolidinones, employed in stereocontrolled aldol and alkylation reactions, synthesis of chiral 2-thiazolidinethiones for aldol reactions, asymmetric alkylation of condensation product with keto acids, asymmetric reactions of its amidines and imines and oxidation of N-protected derivative to the aldehyde.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2026-48-4
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00064296
InChI Key NWYYWIJOWOLJNR-RXMQYKEDSA-N
Synonym l-valinol, s-+-2-amino-3-methyl-1-butanol, 2s-2-amino-3-methylbutan-1-ol, l-+-valinol, h-valinol, s-2-amino-3-methylbutan-1-ol, s-2-amino-3-methyl-1-butanol, 1-butanol, 2-amino-3-methyl-, 2s, s-2-amino-3-methyl-butan-1-ol
PubChem CID 640993
IUPAC Name (2S)-2-amino-3-methylbutan-1-ol
SMILES CC(C)[C@H](N)CO

Specifications

Refractive Index 1.4548
Density 0.926
Boiling Point 80°C to 81°C (8mmHg)
Melting Point 30°C to 35°C
Quantity 5 g
Flash Point 78°C (172°F)
Optical Rotation +17° (c=10 in Ethanol)
Sensitivity Air sensitive
Beilstein 1719137
Solubility Information Soluble in water.
Formula Weight 103.17
Percent Purity 97%
Chemical Name or Material L-(+)-Valinol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-(+)-Valinol, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.