Learn More
OHAUS™ Lab Lift
OHAUS™ Lab Lift provides secure support for holding glassware at elevated positions.
1185.00 DKK - 12745.00 DKK
Product Code | Brand | Material | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Size | Price | Quantity & Availability | |||||
15792419
|
OHAUS™
30400007 |
Aluminum | 102 x 102 (L x W) Platform |
1185.00 DKK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
15702429
|
OHAUS™
30400008 |
Aluminum | 152 x 152 (L x W) Platform |
1455.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15712429
|
OHAUS™
30400009 |
Aluminum | 203 x 203 (L x W) Platform |
1745.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15722429
|
OHAUS™
30400010 |
Aluminum | 254 x 254 (L x W) Platform |
2285.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15722419
|
OHAUS™
30400000 |
Stainless Steel | 76 x 76 (L x W) Platform |
1530.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15732419
|
OHAUS™
30400001 |
Stainless Steel | 102 x 102 (L x W) Platform |
1820.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15742419
|
OHAUS™
30400002 |
Stainless Steel | 152 x 152 (L x W) Platform |
1865.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15752419
|
OHAUS™
30400003 |
Stainless Steel | 203 x 203 (L x W) Platform |
2540.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15762419
|
OHAUS™
30400004 |
Stainless Steel | 254 x 254 (L x W) Platform |
2705.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
15772419
|
OHAUS™
30400005 |
Stainless Steel | 305 x 305 (L x W) Platform |
7195.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
15782419
|
OHAUS™
30400006 |
Stainless Steel | 406 x 406 (L x W) Platform |
12745.00 DKK
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifications
Lab Lift |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
OHAUS™ Lab Lift