missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD2 (LFA-2) Ab-2, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD2 (LFA-2) Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1074-S1

 View more versions of this product

Product Code. 12678856

  • 3940.00 DKK / 500µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CD2, which is also known as E-rosette receptor, T11 and lymphocyte-function antigen-2 (LFA-2), is expressed on T cells, thymocytes, and subset of natural killer cells. Human CD2 functions as the receptor for sheep erythrocytes, human CD58 (LFA-3), and CD15s (Sialyl Lewis X). p56lck, p59fyn, CD3η and CD3ε are tyrosine-phosphorylated after CD2 stimulation. CD2 play a role in T cell activation, T- or NK-cell-mediated cytolysis, apoptosis in activated peripheral T cells, and regulation of T cell anergy.

Host Species: Mouse

Clone: AB75

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Recombinant fragment encoding the external domain of the human CD2 protein

Molecular Weight: 45-58kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
Specifications

Specifications

CD2 (LFA-2) Ab-2
Monoclonal
Unconjugated
Recombinant fragment encoding the external domain of the human CD2 protein
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
AB75
Mouse
500 μL
T-Cell
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.