missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ IgD (δ-Heavy Chain) Ab-1 Rabbit Polyclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ IgD (δ-Heavy Chain) Ab-1, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Brand:  Epredia™ RB-1436-R7

 View more versions of this product

Product Code. 12662907

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: IgD isolated from a pool of normal human sera

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

IgD (delta-Heavy Chain) Ab-1
Polyclonal
Unconjugated
IgD isolated from a pool of normal human sera
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
Polyclonal
Rabbit
7 mL
Immunoglobulins
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.