missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody

Epredia™ MART-1/Melan-A (Melanoma Marker) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia™ MS-799-RQ

 View more versions of this product

Product Code. 12692327

  • 3310.00 DKK / 12mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. MART-1 is present in melanosomes and endoplasmic reticulum.

Host Species: Mouse

Clone: A103

Isotype: IgG1

Species Reactivity: Human and Dog. Others not tested.

Epitope: Not determined

Immunogen: Recombinant MART-1 protein

Molecular Weight: 18kDa

Positive Control: CAC1 melanoma cells and Melanomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)
Specifications

Specifications

MART-1/Melan-A (Melanoma Marker) Ab-4
Monoclonal
Mouse
12 mL
Cancer and Tumor Biology
Ready-to-Use
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
A103
Unconjugated
Recombinant MART-1 protein
IVD
Primary
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.