Learn More
Epredia™ Lab Vision™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-734-S1
2389.20 DKK valid until 2024-12-31
Use promo code "21628" to get your promotional price.
Description
Host Species: Mouse
Clone: 8A9
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Purified human Placental Alkaline Phosphatase (PLAP)
Molecular Weight: 60-70kDa
Positive Control: Placenta or seminoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Placental Alk. Phosphatase (PLAP) Ab-4 | |
Monoclonal | |
Unconjugated | |
Purified human Placental Alkaline Phosphatase (PLAP) | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
8A9 | |
Mouse | |
500 μL | |
Cancer and Tumor Biology | |
Human | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.