Learn More
Labconco™ Ducting Parts: Connectors
Connectors for Labconco ducts
5240.00 DKK
Specifications
For Use With (Equipment) Hoods and Enclosures
View More Specs
Products 1
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
15789688
View Documents
Labconco™
5440900
Connector, Exhaust (Six Thimble)
5240.00 DKK
Each
Description
Description
Powder-coated steel, 6 in. diameter, for connecting the glove box to ductwork attached to a remote blower. Remote blower must be balanced with glove box's built-in blower.
Specifications
Specifications
Hoods and Enclosures