Learn More
Labconco™ Moisture Monitor/Hygrometer
For placement inside the glove box main chamber to continuously monitor moisture levels
Brand:  Labconco™ 5244901
 View more versions of this product
Product Code. 12697755
  • 78700.00 DKK / Each
Only null left
Estimated Shipment: 14-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Measures absolute moisture content in gas
  • Detects levels from 0 to 10,000ppm (with three ranges of 0-100ppm, 100-1000ppm, and 1000-10,000ppm) with ±1 percent accuracy
  • Consists of chemically-inert ceramic technology moisture sensor, sampling block, digital display monitor, sampling pump, switching power supply and on/off switch
  • For use with all glove boxes
Specifications
Specifications
Moisture Monitor/Hygrometer
±1%
18.28 cm
For placement inside the glove box main chamber to continuously monitor moisture levels
5 ppm
7.7 in.
230 V 50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Labconco™ Moisture Monitor/Hygrometer > For 230 volt, 50/60 Hz. operation