Learn More
Labconco™ SpillStopper™ Work Surface for 4 Ft. Protector™ Premier™ Hood
Labconco SpillStopper™ Work Surface is dished and contoured to conform to the interior liner to contain spills.
11225.00 DKK
Specifications
For Use With (Equipment) | Protector XL Laboratory Hood |
---|---|
Product Type | SpillStopper Work Surface (4 ft.) |
Material | Epoxy Resin |
For Use With (Application) | Laboratory Fume Hood Work |
Product Code | Brand | Cut Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cut Style | Price | Quantity & Availability | |||||
16338705
|
Labconco™
9500400 |
No Cutouts | N/A | ||||||
Description
- Corrosion-resistant epoxy resins molded
- Front edge with large radius to aerodynamically direct airflow into hood
- Pre-drilled holes for venting and plumbing pass-throughs provided
- Some surfaces with cutouts for surface-mounted service fixture and/or 6 x 3 in. (15.2 x 7.6cm) cup sinkcup sink
- Flat work surfaces may also be coupled with adjustable height base stand to provide auxiliary workspace
Order Info
Service fixture and cup sink sold separately
Specifications
Protector XL Laboratory Hood | |
SpillStopper Work Surface (4 ft.) | |
Epoxy Resin | |
Laboratory Fume Hood Work |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Labconco™ SpillStopper™ Work Surface for 4 Ft. Protector™ Premier™ Hood