Learn More
Shamrock™ Label Slide
Brand:  Shamrock™ SLR-15RC
278.59 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11832631
  • 420.00 DKK / Pack of 1000
Only null left
Estimated Shipment: 13-08-2025
Specifications
Slide Label
23.8 x 23.8 mm
Roll
White
Litho
1000 Pk.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Shamrock™ Label Slide >