Learn More
Buerkle™ Laboratory Bottle PET Sterile
Sterile laboratory bottle for use in laboratories and chemical handling areas. Good product presentation due to crystal-clear PET. Space-saving and absolutely leakproof. Ideal for storing and packing liquids.
Brand:  Buerkle™ 0370-0125
24.80 DKK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
 View more versions of this product
Product Code. 15722785
  • 26.11 DKK / Each
Elevate Your Beverage Testing with Precision Solutions
Use promo code 24135 at checkout
View Available Promotions
Only null left
Estimated Shipment: 18-07-2025
Description
  • Sterile SAL 10-3 (according to ISO 11137)
  • PET crystal-clear
  • With tamper-proof closure (HDPE)
  • With moulded-in raised graduation
Specifications
Specifications
125 mL
PET
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Buerkle™ Laboratory Bottle PET Sterile > Capacity: 125mL