Learn More
Cytiva (formerly Pall Lab) Laboratory Manifold MicroFunnel™ Adapter
Accessory adapter for MicroFunnel filter funnels.
Brand: Cytiva (formerly Pall Lab) 4890
3225.25 DKK valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- Lab manifold is designed for flexibility and reduction in cross-contamination
- Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
Specifications
Black Fluorocarbon Elastomer O-ring | |
Stainless Steel (316L) | |
Non-sterile | |
3/Pk. |