Learn More
Cytiva Laboratory Manifold Spare O-ring Kit
Replacement complete o-ring kit for microbiology manifold.
Brand:  Cytiva 4878
111.15 DKK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 17164431
  • 117.00 DKK / 1 set
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Description
  • Lab manifold is designed for flexibility and reduction in cross-contamination
  • Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
Non-sterile
1 Set
Fluoroelastomer
11/Pk.
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    23
    Hours
    15
    Minutes
    6
    Seconds
    30
Product Content Correction
Product Title
Cytiva Laboratory Manifold Spare O-ring Kit > Fluorocarbon Elastomer