Learn More
Nickel Electro™ Nickel Crucibles
Get optimum results with nickel crucibles. With working temperature ranges up to 350°C, the Nickel Electro™ Nickel Crucible Capacity consistently has a long working life.
Brand:  Nickel Electro™ 1
 View more versions of this product
Product Code. 11479569
  • 102.00 DKK / Each
Only null left
Estimated Shipment: 08-09-2025
Description
  • Tall form, without lid
  • Suitable for strong alkalis and phosphoric acid
  • Resistant to HF and Cl2below 500°C and to HCl
  • Capacity and dimensions are approximate
  • Working temperature range up to 350°C will ensure crucible has a long working life
  • In the analytical laboratory, nickel crucibles provide a low cost crucible
  • Can be used for research, experiments, chemical analysis, tests, quality control and education
Specifications
Specifications
Nickel
Each
5 mL
25 mm
350°C
0.5 mm
20 mm
Laboratory Crucible
Documents
One moment while we fetch your results.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Nickel Electro™ Nickel Crucibles > Diameter: 20mm; Thickness: 0.5mm