Learn More
Serva Electrophoresis™ Laemmli Sample Buffer, 2X
Brand:  Serva Electrophoresis™ 4252601
Encompass_Preferred
Product Code. 11702177
  • 243.49 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
2X
SDS-PAGE
126mM Tris/Cl (pH 6.8), 20% glycerol, 4% SDS and 0.02% bromophenol blue
20mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications