Learn More
Corning™ Lambda™ EliteTouch™ Pipettor Starter Kit
Corning™ Lambda™ EliteTouch™ Pipettor Starter Kit has been engineered to provide the highest levels of ergonomics, accuracy, and precision.
Brand: Corning™ 6065
5833.20 DKK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Description
Corning Lambda EliteTouch Starter Kit contains 4 variable volume pipettors in the most popular and universal volumes (0.5-10 ÎĽL, 2-20 ÎĽL, 20-200 ÎĽL, and 100-1000 ÎĽL), pipet stand for 4 pipettors, 3 racks with tips, and accessories.
- Cat. No. 6051 Lambda™ EliteTouch™ Single-channel pipettor, 0.5 to 10 μL
- Cat. No. 6052 Lambda EliteTouch Single-channel pipettor, 2 to 20 ÎĽL
- Cat. No. 6055 Lambda EliteTouch Single-channel pipettor, 20 to 200 ÎĽL
- Cat. No. 6056 Lambda EliteTouch Single-channel pipettor, 100 to 1000 ÎĽL
- Cat. No. 6066 Universal linear stand for four single-channel pipettors
- Cat. No.4134 (10 ÎĽL) Corning DeckWorks pipet tips
- Cat. No. 4138 (200 ÎĽL) Corning DeckWorks pipet tips
- Cat. No. 4140 (1000 ÎĽL) Corning DeckWorks pipet tips
- 3 colored pushbuttons (4 sets)
Learn more from Europe, Middle East, and Africa
See resource tab below for specifications.
'
Specifications
ISO8655 | |
Ergonomic | |
Lambda EliteTouch | |
Ergonomic | |
1 |
Autoclavable | |
1 | |
Starter Kit | |
4 single-channel pipettors, universal linear stand for four single-channel pipettors, DeckWorks pipet tips, 3 colored pushbuttons (4 sets), accessories | |
3 Years |
Promotions
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
New Offer Save Up to 78% -
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%