missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Laminated Polyester: ISO Safety Sign - No access for fork lift trucks and other industrial vehicles

Sign compliant to ISO 7010.

25.50 DKK - 57.85 DKK

Specifications

Certifications/Compliance ISO 7010
Finish Gloss
Material Polyester
Adhesive Type Permanent Acrylic
For Use With (Application) Indoor and Outdoor
View More Specs

Products 4
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
15644607
View Documents
Brady™
822590
50 mm
39.35 DKK
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
15654607
View Documents
Brady™
822592
100 mm
25.50 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
15664607
View Documents
Brady™
822593
200 mm
32.80 DKK
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
 
15674607
View Documents
Brady™
822594
315 mm
57.85 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Laminated polyester
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
Polyester
Indoor and Outdoor
-40°C to 110°C
Global
Gloss
Permanent Acrylic
0.099 mm
Round
Black on White
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.