Learn More
Thermo Scientific™ LanthaScreen™ Eu-anti-DYKDDDDK Antibody
LanthaScreen™ Eu-anti-DYKDDDDK Antibody
7190.00 DKK - 85900.00 DKK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10070808
|
Thermo Scientific™
PV6026 |
25 μg |
7190.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10192928
|
Thermo Scientific™
PV6027 |
1 mg |
85900.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LanthaScreen Europium-labeled anti-DYKDDDDK antibodies are used to indirectly label proteins tagged with DYKDDDDK (Flag). This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity.For Research Use Only. Not for use in diagnostic procedures.