Learn More
OHAUS™ Large Three Prong Sleeves
OHAUS™ Large Three Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
Brand:  OHAUS™ 30400262
76.84 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15712679
  • 80.05 DKK / Each
Only null left
Estimated Shipment: 07-07-2025
Description
  • Available in red or black vinyl and fiberglass
  • Fiberglass sleeves are ideal for application above 100°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3
Fiberglass
Gray
Prong Sleeve
Large
Certificates
Promotions
Promotions
Product Content Correction
Product Title
OHAUS™ Large Three Prong Sleeves > Material: Fiberglass; Color: Grey