missing translation for 'onlineSavingsMsg'
Learn More

LATS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen LATS1
Concentration 0.26 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16816874
View Documents
Proteintech
17049-1-AP-20UL
20 ÎĽL
1081.84 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
16806874
View Documents
Proteintech
17049-1-AP-150UL
150 ÎĽL
3058.99 DKK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.
Specifications

Specifications

LATS1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
O95835, Q8BYR2
16798, 308265, 9113
LATS1 Fusion Protein Ag10709
Primary
-20°C
LATS1
0.26 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
h warts, LATS1, WARTS, WARTS protein kinase, wts
LATS1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
LATS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.