Learn More
BRAND™ LDPE Dropping Pipette
Low density polyethylene (LDPE) construction. Used for sampling and decanting infectious or toxic liquids. Graduated with bellows.
294.00 DKK - 331.00 DKK
Specifications
Material | LDPE |
---|---|
For Use With (Application) | For Sampling and Decanting Infectious or Toxic Liquids |
Graduated | Yes |
Quantity | 100/Pk. |
Type | Dropping Pipette |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
10717801
|
BRAND™
125410 |
1.5 mL |
294.00 DKK
Pack of 100 |
In Stock
Log in to see stock. |
||||||
10604161
|
BRAND™
125420 |
5 mL |
331.00 DKK
Pack of 100 |
Estimated Shipment: 27-06-2025
Log in to see stock. |
||||||
Description
- LDPE construction
- Graduated
- Includes bellows
- For handling toxic or infections samples
- Supplied in packs of 100
Specifications
LDPE | |
Yes | |
Dropping Pipette |
For Sampling and Decanting Infectious or Toxic Liquids | |
100/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ LDPE Dropping Pipette