missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ 250 mL Safety Wash Bottles Series 303 LDPE

Designed to give an accurate fine controlled stream of liquid. Kautex™ 250mL Safety Wash Bottles provide excellent chemical resistance for laboratory safety.

Brand:  Kautex™ 2000770020

 View more versions of this product

Product Code. 10657691

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Imprinted with hazard symbols and substances of content in German, English and French language
  • Writing space
  • Abrasion-resistant printing
  • Closure color matches color of ink for easy identification of media
  • Leak-proof
  • Completely emptied by sliding delivery tube, even when inverted.
Suitable for the declared liquids. Not suitable for food.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

250 mL
30.8 mm
10
Red
Round
Wash Bottle
64 mm
Natural
LDPE
150 Pk.
Wide
120 mm
Acetone
Screw Cap with Delivery Nozzle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.