missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ LDPE Lid for Rectangular Container

Buerkle™ LDPE Lid for Rectangular Container is suitable for use with Buerkle™ Rectangular Containers.

Brand:  Buerkle™ 3440-4001

 View more versions of this product

Product Code. 11911471

  • 3532.68 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Linear LDPE
  • Translucent
  • For use with rupture proof rectangular containers
Specifications

Specifications

Container Lid
Rectangular
Lid
50 L Container
320 mm
LDPE
1000 x 320 mm
White
1000 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ LDPE Lid for Rectangular Container > For 50L container

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.