Learn More
Kautex™ LDPE Screw Closure
Feature LDPE light weight, tough construction. Kautex™ LDPE Screw Closures are available in a wide variety of sizes for Kautex™ narrow and wide neck bottles.
Brand: Kautex™ 2000070604-GMP
This item is not returnable.
View return policy
Description
- Made from low density polyethylene
- Translucent
- Available for a wide variety of sizes
Specifications
Natural | |
LDPE | |
50/100/200 mL Narrow Neck Bottle | |
18 mm |
Product Content Correction
Product Title
Kautex™ LDPE Screw Closure > Cap Size: 18 mm O.D.
Spot an opportunity for improvement?Share a Content Correction