Learn More
Mettler Toledo™ Lead Half Cell Electrode
Brand:  Mettler Toledo™ 51107873
9400.81 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11987711
  • 9645.00 DKK / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Specifications
Polymer Membrane
0°C to 50°C
2 to 8
Product Title
Mettler Toledo™ Lead Half Cell Electrode > Membrane: Polymer