Learn More
3M™ Leather Sweatband
3M™ Leather Sweatband with pinlock suspension is designed for use with all 3M™ Hard Hats.
Brand: 3M™ 7000107940
1467.70 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement pinlock suspension cradle with leather sweatband
- Pinlock adjustment system providing simple and secure size adjustment for head sizes from 54 to 62 cm
- Snaps easily into place, to provide all the comfort and easy adjustability of the original system
- 3M™ provides reliable head and face protection products that meet regulated performance and safety requirements
- Suitable for use with all 3M™ Hard Hats
Specifications
Leather | |
Sweatband | |
Pinlock Suspension |
Product Content Correction
Product Title
3M™ Leather Sweatband > Material: Leather
Spot an opportunity for improvement?Share a Content Correction