Learn More
Lecithin, MedChemExpress
MedChemExpress Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells.
Brand:  Medchem Express HY-B2235
Encompass_Preferred
 View more versions of this product
Product Code. 16468876
  • 523.71 DKK / 100mg
Explore more special offers
Description
Description
Lecithin is regarded as a safe, conventional phospholipid source. Phospholipids are reported to alter the fatty acid composition and microstructure of the membranes in animal cells.
Specifications
Specifications
Lecithin
Light Yellow
C42H80NO8P
DMSO : 5 mg/mL (6.60 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (4.39 mM; Need ultrasonic)
758.06
98.0%
Solid
8002-43-5
Research
100 mg
O=P(OC[C@@H](COC(CCCCCCCCCCCCCCC)=O)OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)(OCC[N+](C)(C)C)[O-]
758.06
Research
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Lecithin, MedChemExpress